missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-N-Fmoc-L-phenylalanine, 95%, Thermo Scientific Chemicals

Catalog Number p-7056979
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
198560-44-0
C24H20ClNO4
421.877
MFCD00672548
UOZAKKJRIKXQPY-QFIPXVFZSA-N
fmoc-l-3-chlorophenylalanine, fmoc-3-chloro-l-phenylalanine, fmoc-phe 3-cl-oh, fmoc-l-phe 3-cl-oh, l-3-3-chlorophenyl-n-fmoc-alanine, fmoc-l-3-chlorophe, l-3-chlorophenyl-n-fmoc-alanine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-3-chlorophenyl propanoic acid, 2s-3-3-chlorophenyl-2-9h-fluoren-9-ylmethoxycarbonylamino propanoic acid, 2s-3-3-chlorophenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid
2761466
(2S)-3-(3-chlorophenyl)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC(=CC=C4)Cl)C(=O)O
This item is not returnable. View return policy
198560-44-0
C24H20ClNO4
421.877
MFCD00672548
UOZAKKJRIKXQPY-QFIPXVFZSA-N
fmoc-l-3-chlorophenylalanine, fmoc-3-chloro-l-phenylalanine, fmoc-phe 3-cl-oh, fmoc-l-phe 3-cl-oh, l-3-3-chlorophenyl-n-fmoc-alanine, fmoc-l-3-chlorophe, l-3-chlorophenyl-n-fmoc-alanine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-3-chlorophenyl propanoic acid, 2s-3-3-chlorophenyl-2-9h-fluoren-9-ylmethoxycarbonylamino propanoic acid, 2s-3-3-chlorophenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid
2761466
(2S)-3-(3-chlorophenyl)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC(=CC=C4)Cl)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Formula Weight 421.88
Percent Purity 95%
Quantity 1 g
Physical Form Solid
Chemical Name or Material 3-Chloro-N-Fmoc-L-phenylalanine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'