Alfa Aesar™ 3-Chloro-N-(2,4-difluorophenyl)benzamide, 97%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: H58231.ME

UNSPSC: 12352106

Code: NEW

Additional Details:
Additional Details: CAS Number: 331435-75-7 Weight: 0.00050kg


Quantity 500mg

Product Code. 15434558

Quantity Price
1 £ 223.55 / 500mg
Estimated Shipment date
from Supplier 11-04-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 3-Chloro-N-(2,4-difluorophenyl)benzamide
CAS 331435-75-7
Formula Weight 267.66
MDL Number MFCD00751584

Specification

Formula Weight 267.66
Physical Form Solid
CAS 331435-75-7
Quantity 500mg
Assay 97%
MDL Number MFCD00751584
Chemical Name or Material 3-Chloro-N-(2,4-difluorophenyl)benzamide
Documentation

Technical documents