missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-D-phenylalanine, 98%, Thermo Scientific™

Catalog Number 15419838
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
80126-52-9
C9H10ClNO2
199.634
MFCD01631984
JJDJLFDGCUYZMN-MRVPVSSYSA-N
3-chloro-d-phenylalanine, h-d-phe 3-cl-oh, r-2-amino-3-3-chlorophenyl propanoic acid, d-3-chlorophenylalanine, 2r-2-amino-3-3-chlorophenyl propanoic acid, d-phe 3-cl-oh, d-3-chlorophe, 3-chloro-d-phe-oh, d-phenylalanine, 3-chloro, pubchem11977
7009409
(2R)-2-amino-3-(3-chlorophenyl)propanoic acid
C1=CC(=CC(=C1)Cl)CC(C(=O)O)N
This item is not returnable. View return policy
80126-52-9
C9H10ClNO2
199.634
MFCD01631984
JJDJLFDGCUYZMN-MRVPVSSYSA-N
3-chloro-d-phenylalanine, h-d-phe 3-cl-oh, r-2-amino-3-3-chlorophenyl propanoic acid, d-3-chlorophenylalanine, 2r-2-amino-3-3-chlorophenyl propanoic acid, d-phe 3-cl-oh, d-3-chlorophe, 3-chloro-d-phe-oh, d-phenylalanine, 3-chloro, pubchem11977
7009409
(2R)-2-amino-3-(3-chlorophenyl)propanoic acid
C1=CC(=CC(=C1)Cl)CC(C(=O)O)N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color White
Formula Weight 199.63
Percent Purity 98%
Quantity 250 mg
Physical Form Powder
Chemical Name or Material 3-Chloro-D-phenylalanine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'