missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-5-(trifluoromethyl)phenylboronic acid, 95%, ACROS Organics™

Brand:  Acros Organics 445370010

UNSPSC : 12161500

Code : 29

Additional Details : CAS Number : 1160561-31-8 Weight : 0.05100kg


Quantity 1g

 View more versions of this product

Product Code. 11956131

  • £50.00 / 1g


Estimated Shipment
13-09-2017
Add to basket


Specifications

Chemical Identifiers

3-Chloro-5-(trifluoromethyl)phenylboronic acid
95%
C7H5BClF3O2
224.38

Specifications

3-Chloro-5-(trifluoromethyl)phenylboronic acid
1g
95%
C7H5BClF3O2
210.0° to 212.0°C
1160561-31-8
95%
224.38
Safety and Handling

Safety and Handling

Product Identifier
  • 3-Chloro-5-(trifluoromethyl)phenylboronic acid

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

Documentation

Technical Documents