missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-5-(trifluoromethyl)phenylboronic acid, 95%, ACROS Organics™

Brand:  Acros Organics 445370050

Code : 29

Additional Details : CAS Number : 1160561-31-8 Weight : 0.05500kg


Quantity 5g

 View more versions of this product

Product Code. 11966131

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

3-Chloro-5-(trifluoromethyl)phenylboronic acid
95%
C7H5BClF3O2
224.38

Specifications

3-Chloro-5-(trifluoromethyl)phenylboronic acid
1160561-31-8
C7H5BClF3O2
5g
95%
224.38
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 3-Chloro-5-(trifluoromethyl)phenylboronic acid

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS