missing translation for 'onlineSavingsMsg'
Learn More

3-chloro-5-fluorobenzyl Bromide, 97%, Thermo Scientific™

Catalog Number 10727533
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
493024-39-8
C7H5BrClF
223.47
MFCD03788472
PKSAHOPFPPAUOD-UHFFFAOYSA-N
3-chloro-5-fluorobenzyl bromide, 1-bromomethyl-3-chloro-5-fluorobenzene, 3-chloro-5-fluorobenzylbromide, 5-chloro-3-fluorobenzyl bromide, benzene, 1-bromomethyl-3-chloro-5-fluoro, 5-bromomethyl-3-chloro-1-fluorobenzene, alpha-bromo-3-chloro-5-fluorotoluene, pubchem4896, acmc-209kfr, 1-bromomethyl-3-chloro-5-fluoro-benzene
2734834
1-(bromomethyl)-3-chloro-5-fluorobenzene
FC1=CC(Cl)=CC(CBr)=C1
This item is not returnable. View return policy
493024-39-8
C7H5BrClF
223.47
MFCD03788472
PKSAHOPFPPAUOD-UHFFFAOYSA-N
3-chloro-5-fluorobenzyl bromide, 1-bromomethyl-3-chloro-5-fluorobenzene, 3-chloro-5-fluorobenzylbromide, 5-chloro-3-fluorobenzyl bromide, benzene, 1-bromomethyl-3-chloro-5-fluoro, 5-bromomethyl-3-chloro-1-fluorobenzene, alpha-bromo-3-chloro-5-fluorotoluene, pubchem4896, acmc-209kfr, 1-bromomethyl-3-chloro-5-fluoro-benzene
2734834
1-(bromomethyl)-3-chloro-5-fluorobenzene
FC1=CC(Cl)=CC(CBr)=C1
CAS Min % 96.0
CAS Max % 100.0
Melting Point 25°C to 30°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula ClC6H3(F)CH2Br
Quantity 1 g
Formula Weight 223.47
Percent Purity 97%
Physical Form Low Melting Crystals, Crystalline Mass or Liquid After Melting
Chemical Name or Material 3-Chloro-5-fluorobenzyl bromide
Show More Show Less
compliance-icons
Product Identifier
  • 3-Chloro-5-fluorobenzyl bromide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'