missing translation for 'onlineSavingsMsg'
Learn More

3-chloro-5-fluoroaniline, 97%, ACROS Organics™

Brand:  Acros Organics 374740250

Code : 29

Additional Details : CAS Number : 4863-91-6 Weight : 0.07500kg Transport : UN number : 2810 Chem class : 1 Pack group : III


Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10696964

  • £857.00 / 25g


EU Stock 2
Add to basket


Specifications

Chemical Identifiers

3-chloro-5-fluoroaniline
96.0
≥96.0%
C6H5ClFN
145.56
4863-91-6
100.0
97%
ClC6H3(F)NH2
MFCD03407962

Specifications

3-chloro-5-fluoroaniline
Clear light yellow to orange
4863-91-6
100.0
Glass bottle
Darkens on exposure to light and air
≥96.0%
C6H5ClFN
145.56
1.45g/mL
25g
96.0
Authentic
1.5610 to 1.5640 (20°C, 589nm)
Liquid
97%
ClC6H3(F)NH2
MFCD03407962
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 3-Chloro-5-fluoroaniline

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS