missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-(trifluoromethyl)benzoyl chloride, 97%, Alfa Aesar™

Brand:  Alfa Aesar H32343.03

Code : NEW

Additional Details : Weight : 0.00100kg Transport : UN number : 3265 Chem class : 8 Pack group : II

Quantity 1g

 View more versions of this product

Product Code. 15485057

  • £54.90 / 1g


Estimated Shipment date
from Supplier 31-05-2018
Add to basket

Specifications

Chemical Identifiers

3-Chloro-4-(trifluoromethyl)benzoyl chloride
243.01
UN3265
C8H3Cl2F3O
MFCD11519325

Specifications

3-Chloro-4-(trifluoromethyl)benzoyl chloride
Colorless
1092460-77-9
Liquid
C8H3Cl2F3O
MFCD11519325
Moisture Sensitive
1g
1.5030
97%
243.01
UN3265
Videos
SDS