missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3'-Chloro-4'-(trifluoromethoxy)acetophenone, 98%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
3′-Chloro-4′-(trifluoromethoxy)acetophenone is used as an organic chemical synthesis intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
3′-Chloro-4′-(trifluoromethoxy)acetophenone is used as an organic chemical synthesis intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Color | Colorless |
Quantity | 1 g |
Solubility Information | Slightly soluble in water. |
Formula Weight | 238.59 |
Percent Purity | 98% |
Physical Form | Liquid |
Chemical Name or Material | 3'-Chloro-4'-(trifluoromethoxy)acetophenone |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'