missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-nitrotoluene, 98%, ACROS Organics™

Brand:  Acros Organics 348080050

Code : 29

Additional Details : CAS Number : 38939-88-7 Weight : 0.05500kg Transport : UN number : 3457 Chem class : 1 Pack group : III


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10254891

  • £22.20 / 5g


EU Stock 2
Add to basket


Specifications

Chemical Identifiers

3-Chloro-4-nitrotoluene
97.5
98%
C7H6ClNO2
171.58
38939-88-7
100.0
98%
CH3C6H3(NO2)Cl
MFCD02683043

Specifications

3-Chloro-4-nitrotoluene
108.0°C (3.0 mmHg)
22.0°C
38939-88-7
100.0
Glass bottle
Liquid After Melting
98%
CH3C6H3(NO2)Cl
MFCD02683043
Solubility in water: insoluble.
>230°C
Clear yellow
5g
97.5
Authentic
1.5610 to 1.5640
98%
C7H6ClNO2
171.58
05, IV, 856
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 3-Chloro-4-nitrotoluene

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS