missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-nitrotoluene, 98%, ACROS Organics™

Brand:  Acros Organics 348080250

Code : 29

Additional Details : CAS Number : 38939-88-7 Weight : 0.08000kg Transport : UN number : 3457 Chem class : 1 Pack group : III


Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10071871

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

3-Chloro-4-nitrotoluene
97.5
98%
C7H6ClNO2
171.58
38939-88-7
100.0
98%
CH3C6H3(NO2)Cl
MFCD02683043

Specifications

3-Chloro-4-nitrotoluene
108.0°C (3.0 mmHg)
22.0°C
38939-88-7
100.0
Glass bottle
Liquid After Melting
98%
CH3C6H3(NO2)Cl
MFCD02683043
Solubility in water: insoluble.
>230°C
Clear yellow
25g
97.5
Authentic
1.5610 to 1.5640
98%
C7H6ClNO2
171.58
05, IV, 856
Videos
SDS