missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-methylpyridine, 95%, Thermo Scientific™

Catalog Number p-7053881
Click to view available options
Quantity:
5 g
This item is not returnable. View return policy
72093-04-0
C6H6ClN
127.57
MFCD04114245
VLNXNJMHLZFECV-UHFFFAOYSA-N
3-chloro-4-picoline, pyridine, 3-chloro-4-methyl, 3-chloro-4-methyl-pyridine, pubchem14323, acmc-1bl3c, 3-chloro-4-methylpyridine?, ksc494c8t, 3-chloro-4-methylpyridine
2762925
3-chloro-4-methylpyridine
CC1=C(Cl)C=NC=C1
This item is not returnable. View return policy
72093-04-0
C6H6ClN
127.57
MFCD04114245
VLNXNJMHLZFECV-UHFFFAOYSA-N
3-chloro-4-picoline, pyridine, 3-chloro-4-methyl, 3-chloro-4-methyl-pyridine, pubchem14323, acmc-1bl3c, 3-chloro-4-methylpyridine?, ksc494c8t, 3-chloro-4-methylpyridine
2762925
3-chloro-4-methylpyridine
CC1=C(Cl)C=NC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Colorless to Brown
Quantity 5 g
Formula Weight 127.57
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 3-Chloro-4-methylpyridine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'