missing translation for 'onlineSavingsMsg'
Learn More

3-chloro-4-methylbenzoic acid, 97%, Thermo Scientific™

Catalog Number 10170752
Click to view available options
Quantity:
1 g
5 g
10 g
This item is not returnable. View return policy
5162-82-3
C8H7ClO2
170.592
MFCD00045841
SDKUOEOJAXGCLU-UHFFFAOYSA-N
3-chloro-p-toluic acid, p-toluic acid, 3-chloro, benzoic acid, 3-chloro-4-methyl, 3-chloro-4-methyl-benzoic acid, 3-chloro-4-methyl benzoic acid, pubchem4569, acmc-1ayqf, 4-09-00-01743 beilstein handbook reference, ksc272s4h, 3-chloro,4-methylbenzoic acid
78840
3-chloro-4-methylbenzoic acid
CC1=C(C=C(C=C1)C(=O)O)Cl
This item is not returnable. View return policy
5162-82-3
C8H7ClO2
170.592
MFCD00045841
SDKUOEOJAXGCLU-UHFFFAOYSA-N
3-chloro-p-toluic acid, p-toluic acid, 3-chloro, benzoic acid, 3-chloro-4-methyl, 3-chloro-4-methyl-benzoic acid, 3-chloro-4-methyl benzoic acid, pubchem4569, acmc-1ayqf, 4-09-00-01743 beilstein handbook reference, ksc272s4h, 3-chloro,4-methylbenzoic acid
78840
3-chloro-4-methylbenzoic acid
CC1=C(C=C(C=C1)C(=O)O)Cl
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 170.59
Percent Purity 97%
Chemical Name or Material 3-chloro-4-methylbenzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'