missing translation for 'onlineSavingsMsg'
Learn More

3-chloro-4-methylbenzaldehyde, 97%, ACROS Organics™

Brand:  Acros Organics 436690010

Code : 29

Additional Details : CAS Number : 3411-03-8 Weight : 0.05100kg


Quantity 1g

 View more versions of this product

Product Code. 10812811

  • £31.60 / 1g


Estimated Shipment
15-02-2018
Add to basket


Specifications

Chemical Identifiers

3-chloro-4-methylbenzaldehyde
100.0
97%
154.6
96.0
≥96.0%
C8H7ClO

Specifications

3-chloro-4-methylbenzaldehyde
100.0
Low Melting Solid or Liquid
White or colorless
1g
154.6
96.0
Authentic
≥96.0%
97%
C8H7ClO
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 3-Chloro-4-methylbenzaldehyde

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS