missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Chloro-4-methoxybenzylamine hydrochloride, 97%

Brand:  Alfa Aesar™ H63806.06

Code : NEW

Additional Details : CAS Number : 41965-95-1 Weight : 0.00500kg Transport : UN number : 2811 Chem class : 1 Pack group : III

Quantity 5g

 View more versions of this product

Product Code. 15420699

  • £66.40 / 5g


Estimated Shipment date
from Supplier 29-05-2018
Add to basket

Specifications

Chemical Identifiers

3-Chloro-4-methoxybenzylamine hydrochloride
208.09
UN2811
41965-95-1
MFCD01211460

Specifications

3-Chloro-4-methoxybenzylamine hydrochloride
5g
Solid
208.09
UN2811
250° to 255°C
41965-95-1
97%
MFCD01211460
Videos
SDS