missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Chloro-4-hydroxybenzoic acid, 97%, may contain up to ≈6% water

Brand:  Alfa Aesar™ A12134.09

Code : NEW

Additional Details : CAS Number : 3964-58-7 Weight : 0.01000kg

Quantity 10g

 View more versions of this product

Product Code. 11410093

  • £21.00 / 10g


Estimated Shipment date
from Supplier 10-05-2018
Add to basket

Specifications

Chemical Identifiers

3-Chloro-4-hydroxybenzoic acid
C7H5ClO3
MFCD00002549
3964-58-7
172.57
2207866

Specifications

3-Chloro-4-hydroxybenzoic acid
168° to 170°C
3964-58-7
C7H5ClO3
MFCD00002549
May contain up to ≈6% water
10g
97%
172.57
2207866
Videos
SDS