missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-hydroxy-5-methoxyphenylboronic acid pinacol ester, 97%, Thermo Scientific™

Catalog Number 11976061
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1003298-84-7
C13H18BClO4
284.55
XBEJPOZGAPQFPL-UHFFFAOYSA-N
2-chloro-6-methoxy-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 3-chloro-4-hydroxy-5-methoxyphenylboronic acid, pinacol ester, 3-chloro-4-hydroxy-5-methoxyphenylboronic acid pinacol ester, 2-chloro-6-methoxy-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 6-chloro-2-methoxy-4-4,4,5,5-tetra methyl-1,3,2-dioxaborolan-2-yl phenol, phenol, 2-chloro-6-methoxy-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
17750230
2-chloro-6-methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C(=C2)Cl)O)OC
This item is not returnable. View return policy
1003298-84-7
C13H18BClO4
284.55
XBEJPOZGAPQFPL-UHFFFAOYSA-N
2-chloro-6-methoxy-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 3-chloro-4-hydroxy-5-methoxyphenylboronic acid, pinacol ester, 3-chloro-4-hydroxy-5-methoxyphenylboronic acid pinacol ester, 2-chloro-6-methoxy-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 6-chloro-2-methoxy-4-4,4,5,5-tetra methyl-1,3,2-dioxaborolan-2-yl phenol, phenol, 2-chloro-6-methoxy-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
17750230
2-chloro-6-methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C(=C2)Cl)O)OC
Melting Point 156°C to 159°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 284.55
Percent Purity 97%
Chemical Name or Material 3-Chloro-4-hydroxy-5-methoxyphenylboronic acid pinacol ester
compliance-icons
Product Identifier
  • 3-Chloro-4-hydroxy-5-methoxyphenylboronic acid pinacol ester
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'