missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Chloro-4-fluorotoluene, 98%

Brand:  Alfa Aesar™ H63822.14

UNSPSC : 12352101

Code : NEW

Additional Details : CAS Number : 1513-25-3 Weight : 0.02500kg Transport : UN number : 1993 Chem class : 1 Pack group : III

Quantity 25g

 View more versions of this product

Product Code. 15420729

  • £71.97 / 25g


Estimated Shipment date
from Supplier 10-07-2017
Add to basket

Specifications Description & Specifications

Chemical Identifiers

3-Chloro-4-fluorotoluene
144.57
UN1993
1513-25-3
MFCD00079730
2-Chloro-1-fluoro-4-methylbenzene

Specifications

3-Chloro-4-fluorotoluene
167° to 169°C
1513-25-3
98%
MFCD00079730
2-Chloro-1-fluoro-4-methylbenzene
1.18g/mL
25g
Liquid
144.57
UN1993
Documentation

Technical Documents