missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-fluorothiophenol, 97%, Thermo Scientific Chemicals

Catalog Number p-7081050
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
60811-23-6
C6H3ClFS
161.60
MFCD00052938
SFSHSIFYTWIGSF-UHFFFAOYSA-M
3-chloro-4-fluorothiophenol, 3-chloro-4-fluorobenzene-1-thiol, 3-chloro-4-fluoro-benzenethiol, pubchem2857, acmc-209wz2, 3-chloro-4-flurothiophenol, benzenethiol, 3-chloro-4-fluoro
2734197
3-chloro-4-fluorobenzenethiol
FC1=CC=C([S-])C=C1Cl
This item is not returnable. View return policy
60811-23-6
C6H3ClFS
161.60
MFCD00052938
SFSHSIFYTWIGSF-UHFFFAOYSA-M
3-chloro-4-fluorothiophenol, 3-chloro-4-fluorobenzene-1-thiol, 3-chloro-4-fluoro-benzenethiol, pubchem2857, acmc-209wz2, 3-chloro-4-flurothiophenol, benzenethiol, 3-chloro-4-fluoro
2734197
3-chloro-4-fluorobenzenethiol
FC1=CC=C([S-])C=C1Cl

Used as pharamaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as pharamaceutical intermediate.

Solubility
Sparingly Soluble (0.014 g/L) (25°C).

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
TRUSTED_SUSTAINABILITY
Boiling Point 68°C to 70°C (2 mmHg)
Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Sparingly Soluble (0.014g/L) (25°C).
Formula Weight 162.61
Percent Purity 97%
Chemical Name or Material 3-Chloro-4-fluorothiophenol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'