missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Chloro-4-fluorobenzoyl chloride, 97%

£59.02 - £166.24

Chemical Identifiers

CAS 65055-17-6
Molecular Formula C7H3Cl2FO
Molecular Weight (g/mol) 192.998
MDL Number MFCD00236215
InChI Key DLZUHSFUBZTBQZ-UHFFFAOYSA-N
Synonym 4-fluoro-3-chlorobenzoyl chloride, 3-chloro-4-fluorobenzoylchloride, 3-chloro-4-fluoro benzoyl chloride, 3-chloro-4-fluoro-benzoyl chloride, benzoyl chloride, 3-chloro-4-fluoro, acmc-209np6, dsstox_cid_29301, dsstox_rid_83419, dsstox_gsid_49344, ksc622a8f
PubChem CID 2736548
IUPAC Name 3-chloro-4-fluorobenzoyl chloride
SMILES C1=CC(=C(C=C1C(=O)Cl)Cl)F
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11409676
SDS View Documents
Alfa Aesar™
B20217.06
5g £59.02
5g View alternative products
Save Up to 
»
Estimated Shipment
08-10-2020
Please to purchase this item. Need a web account? Register with us today!
11419676
SDS View Documents
Alfa Aesar™
B20217.14
25g £166.24
25g View alternative products
Save Up to 
»
Estimated Shipment
08-10-2020
Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

65055-17-6
192.998
DLZUHSFUBZTBQZ-UHFFFAOYSA-N
2736548
C1=CC(=C(C=C1C(=O)Cl)Cl)F
C7H3Cl2FO
MFCD00236215
4-fluoro-3-chlorobenzoyl chloride, 3-chloro-4-fluorobenzoylchloride, 3-chloro-4-fluoro benzoyl chloride, 3-chloro-4-fluoro-benzoyl chloride, benzoyl chloride, 3-chloro-4-fluoro, acmc-209np6, dsstox_cid_29301, dsstox_rid_83419, dsstox_gsid_49344, ksc622a8f
3-chloro-4-fluorobenzoyl chloride

Specifications

3-Chloro-4-fluorobenzoyl chloride
Moisture sensitive
5g
C7H3Cl2FO
UN3265
4-fluoro-3-chlorobenzoyl chloride, 3-chloro-4-fluorobenzoylchloride, 3-chloro-4-fluoro benzoyl chloride, 3-chloro-4-fluoro-benzoyl chloride, benzoyl chloride, 3-chloro-4-fluoro, acmc-209np6, dsstox_cid_29301, dsstox_rid_83419, dsstox_gsid_49344, ksc622a8f
C1=CC(=C(C=C1C(=O)Cl)Cl)F
192.998
193.01
1.551
222°C
65055-17-6
MFCD00236215
6288904
DLZUHSFUBZTBQZ-UHFFFAOYSA-N
3-chloro-4-fluorobenzoyl chloride
2736548
97%