missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Chloro-4-fluorobenzeneboronic acid pinacol ester, 96%

Brand:  Alfa Aesar™ H62307.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15467266

  • £302.30 / 1g


Estimated Shipment date
from Supplier 08-05-2018
Add to basket

Specifications

Chemical Identifiers

3-Chloro-4-fluorobenzeneboronic acid pinacol ester
256.51
3-Chloro-4-fluorophenylboronic acid pinacol ester

Specifications

3-Chloro-4-fluorobenzeneboronic acid pinacol ester
Solid
256.51
1g
96%
3-Chloro-4-fluorophenylboronic acid pinacol ester
Videos
SDS