missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-fluorobenzene-1-thiol, Tech., Thermo Scientific™

Catalog Number p-2699828
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
60811-23-6
C6H3ClFS
161.60
MFCD00052938
SFSHSIFYTWIGSF-UHFFFAOYSA-M
3-chloro-4-fluorothiophenol, 3-chloro-4-fluorobenzene-1-thiol, 3-chloro-4-fluoro-benzenethiol, pubchem2857, acmc-209wz2, 3-chloro-4-flurothiophenol, benzenethiol, 3-chloro-4-fluoro
2734197
3-chloro-4-fluorobenzenethiol
FC1=CC=C([S-])C=C1Cl
This item is not returnable. View return policy
60811-23-6
C6H3ClFS
161.60
MFCD00052938
SFSHSIFYTWIGSF-UHFFFAOYSA-M
3-chloro-4-fluorothiophenol, 3-chloro-4-fluorobenzene-1-thiol, 3-chloro-4-fluoro-benzenethiol, pubchem2857, acmc-209wz2, 3-chloro-4-flurothiophenol, benzenethiol, 3-chloro-4-fluoro
2734197
3-chloro-4-fluorobenzenethiol
FC1=CC=C([S-])C=C1Cl
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 162.61
Percent Purity Tech.
Grade Technical
Chemical Name or Material 3-Chloro-4-fluorobenzene-1-thiol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'