missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-fluoroaniline, 98%, Thermo Scientific Chemicals

Catalog Number p-87472
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
367-21-5
C6H5ClFN
145.56
MFCD00007767
YSEMCVGMNUUNRK-UHFFFAOYSA-N
benzenamine, 3-chloro-4-fluoro, 3-chloro-4-fluorobenzenamine, 3-chloro-4-fluoro-phenylamine, aniline, 3-chloro-4-fluoro, 3-chloro-4-fluoro aniline, unii-7l63cc70uq, 3-chloro-4-fluorophenylamine, 4-fluoro-3-chloroaniline, 3-chloro-4-fluoro-aniline, 3-chlor-4-fluoranilin
9708
3-chloro-4-fluoroaniline
NC1=CC=C(F)C(Cl)=C1
This item is not returnable. View return policy
367-21-5
C6H5ClFN
145.56
MFCD00007767
YSEMCVGMNUUNRK-UHFFFAOYSA-N
benzenamine, 3-chloro-4-fluoro, 3-chloro-4-fluorobenzenamine, 3-chloro-4-fluoro-phenylamine, aniline, 3-chloro-4-fluoro, 3-chloro-4-fluoro aniline, unii-7l63cc70uq, 3-chloro-4-fluorophenylamine, 4-fluoro-3-chloroaniline, 3-chloro-4-fluoro-aniline, 3-chlor-4-fluoranilin
9708
3-chloro-4-fluoroaniline
NC1=CC=C(F)C(Cl)=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 42°C to 47°C
CAS Min % 97.5
Color Beige to Brown
CAS Max % 100.0
Boiling Point 230°C
Quantity 25 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Solid
Assay Percent Range 98%
Linear Formula ClC6H3(F)NH2
Beilstein 12, I, 309
Solubility Information Solubility in water: 10g/L water 520°C). Other solubilities: soluble in organic solvents
Formula Weight 145.56
Percent Purity 98%
Chemical Name or Material 3-Chloro-4-fluoroaniline, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Chloro-4-fluoroaniline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'