missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Chloro-4-ethoxy-2-fluorobenzeneboronic acid, 98%

Brand:  Alfa Aesar™ H64675.03

Code : NEW

Additional Details : CAS Number : 909122-50-5 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15471979

  • £23.96 / 1g


Estimated Shipment date
from Supplier 09-07-2018
Add to basket

Specifications

Chemical Identifiers

3-Chloro-4-ethoxy-2-fluorobenzeneboronic acid
218.42
14609288
909122-50-5
MFCD15144702
3-Chloro-4-ethoxy-2-fluorophenylboronic acid

Specifications

3-Chloro-4-ethoxy-2-fluorobenzeneboronic acid
909122-50-5
98%
MFCD15144702
3-Chloro-4-ethoxy-2-fluorophenylboronic acid
1g
Solid
218.42
14609288
Videos
SDS