missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-nitrobenzoic acid, 98%, ACROS Organics™

Brand:  Acros Organics 151690050

Code : 29

Additional Details : CAS Number : 4771-47-5 Weight : 0.05500kg


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10377891

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

3-Chloro-2-nitrobenzoic acid
97.5
≥97.5%
C7H4ClNO4
201.57
4771-47-5
100.0
98%
ClC6H3(NO2)CO2H
MFCD00007087

Specifications

3-Chloro-2-nitrobenzoic acid
97.5
100.0
236° to 240°C
Almost white to beige
≥97.5%
C7H4ClNO4
201.57
4771-47-5
Authentic
Glass bottle
5g
Crystalline Powder
98%
ClC6H3(NO2)CO2H
MFCD00007087
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 3-Chloro-2-nitrobenzoic acid

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS