missing translation for 'onlineSavingsMsg'
Learn More

3-chloro-2-methylphenyl isothiocyanate, 97%, Thermo Scientific™

Catalog Number 10643043
Click to view available options
Quantity:
1 g
10 g
25 g
This item is not returnable. View return policy
19241-35-1
C8H6ClNS
183.653
MFCD00022056
ZXEZATIRZLJXFU-UHFFFAOYSA-N
140504
1-chloro-3-isothiocyanato-2-methylbenzene
CC1=C(C=CC=C1Cl)N=C=S
This item is not returnable. View return policy
19241-35-1
C8H6ClNS
183.653
MFCD00022056
ZXEZATIRZLJXFU-UHFFFAOYSA-N
140504
1-chloro-3-isothiocyanato-2-methylbenzene
CC1=C(C=CC=C1Cl)N=C=S
CAS Min % 97.0
Assay Percent Range 97%
Synonym 3-chloro-2-methylphenyl isothiocyanate, isothiocyanic acid, 3-chloro-o-tolyl ester, 3-chloro-2-methylphenylisothiocyanate, benzene,1-chloro-3-isothiocyanato-2-methyl, 3-chloro-2-methylbenzenisothiocyanate, pubchem10477, 3-chloro-2-methylphenyl-isothiocyanate, 1-chloranyl-3-isothiocyanato-2-methyl-benzene, benzene, 1-chloro-3-isothiocyanato-2-methyl
Formula Weight 183.66
Percent Purity 97%
Quantity 1 g
Packaging Amber Glass Bottle
Chemical Name or Material 3-Chloro-2-methylphenyl isothiocyanate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'