missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-methylbenzoic acid, 98%, Thermo Scientific™

Catalog Number 15422212
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
7499-08-3
C8H7ClO2
170.6
HXGHMCLCSPQMOR-UHFFFAOYSA-N
3-chloro-o-toluic acid, 3-chloro-2-methyl-benzoic acid, benzoic acid, 3-chloro-2-methyl, o-toluic acid, 3-chloro, 3-chloro-2-methylbenzoicacid, 2-methyl-3-chlorobenzoic acid, pubchem8362, 2-carboxy-6-chlorotoluene, 3chloro-2-methylbenzoic acid, 3-chloro-2-methybenzoic acid
82010
3-chloro-2-methylbenzoic acid
CC1=C(C=CC=C1Cl)C(=O)O
This item is not returnable. View return policy
7499-08-3
C8H7ClO2
170.6
HXGHMCLCSPQMOR-UHFFFAOYSA-N
3-chloro-o-toluic acid, 3-chloro-2-methyl-benzoic acid, benzoic acid, 3-chloro-2-methyl, o-toluic acid, 3-chloro, 3-chloro-2-methylbenzoicacid, 2-methyl-3-chlorobenzoic acid, pubchem8362, 2-carboxy-6-chlorotoluene, 3chloro-2-methylbenzoic acid, 3-chloro-2-methybenzoic acid
82010
3-chloro-2-methylbenzoic acid
CC1=C(C=CC=C1Cl)C(=O)O
Melting Point 163.0°C to 166.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Formula Weight 170.6
Percent Purity 98%
Chemical Name or Material 3-Chloro-2-methylbenzoic acid
compliance-icons
Product Identifier
  • 3-Chloro-2-methylbenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'