missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-methylaniline 99%, Thermo Scientific Chemicals

Catalog Number p-88632
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
87-60-5
C7H8ClN
141.60
MFCD00007766
ZUVPLKVDZNDZCM-UHFFFAOYSA-N
3-chloro-o-toluidine, 2-amino-6-chlorotoluene, benzenamine, 3-chloro-2-methyl, 3-chloro-2-toluidine, scarlet tr base, fast scarlet tr base, 6-chloro-2-aminotoluene, o-toluidine, 3-chloro, 3-chloro-2-methylphenylamine, o-methyl-m-chloroaniline
6894
3-chloro-2-methylaniline
CC1=C(N)C=CC=C1Cl
This item is not returnable. View return policy
87-60-5
C7H8ClN
141.60
MFCD00007766
ZUVPLKVDZNDZCM-UHFFFAOYSA-N
3-chloro-o-toluidine, 2-amino-6-chlorotoluene, benzenamine, 3-chloro-2-methyl, 3-chloro-2-toluidine, scarlet tr base, fast scarlet tr base, 6-chloro-2-aminotoluene, o-toluidine, 3-chloro, 3-chloro-2-methylphenylamine, o-methyl-m-chloroaniline
6894
3-chloro-2-methylaniline
CC1=C(N)C=CC=C1Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 2.0°C
Color Brown-Red or Red to Yellow
Density 1.1710g/mL
Boiling Point 245.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula ClC6H3(CH3)NH2
Refractive Index 1.5870 to 1.5890
Quantity 100 mL
Beilstein 12, 836
Formula Weight 141.6
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3-Chloro-2-methylaniline, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Chloro-2-methylaniline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'