missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-methoxypyridine-4-boronic acid, 95%, Thermo Scientific™

Catalog Number 13488618
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
957060-88-7
C6H7BClNO3
187.39
MFCD06798255
YQNAXOGPSUVHNU-UHFFFAOYSA-N
3-chloro-2-methoxypyridine-4-boronic acid, 3-chloro-2-methoxypyridin-4-yl boronic acid, 3-chloro-2-methoxypyridin-4-ylboronicacid, 2-methoxy-3-chloropyridine-4-boronic acid, 3-chloro-2-methoxy-4-pyridineboronic acid, pubchem17058, abbypharma ap-17-5909, 3-chloro-2-methoxypyrdine-4-boronic acid, boronic acid, b-3-chloro-2-methoxy-4-pyridinyl
44119809
(3-chloro-2-methoxypyridin-4-yl)boronic acid
COC1=NC=CC(B(O)O)=C1Cl
This item is not returnable. View return policy
957060-88-7
C6H7BClNO3
187.39
MFCD06798255
YQNAXOGPSUVHNU-UHFFFAOYSA-N
3-chloro-2-methoxypyridine-4-boronic acid, 3-chloro-2-methoxypyridin-4-yl boronic acid, 3-chloro-2-methoxypyridin-4-ylboronicacid, 2-methoxy-3-chloropyridine-4-boronic acid, 3-chloro-2-methoxy-4-pyridineboronic acid, pubchem17058, abbypharma ap-17-5909, 3-chloro-2-methoxypyrdine-4-boronic acid, boronic acid, b-3-chloro-2-methoxy-4-pyridinyl
44119809
(3-chloro-2-methoxypyridin-4-yl)boronic acid
COC1=NC=CC(B(O)O)=C1Cl
Melting Point 140°C
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 250 mg
Formula Weight 187.39
Percent Purity 95%
Chemical Name or Material 3-Chloro-2-methoxypyridine-4-boronic acid
compliance-icons
Product Identifier
  • 3-Chloro-2-methoxypyridine-4-boronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'