missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-fluoro-5-(trifluoromethyl)pyridine, 98%, Thermo Scientific™

Catalog Number p-7055993
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
72537-17-8
C6H2ClF4N
199.53
MFCD00191918
GDSROTVTTLUHCO-UHFFFAOYSA-N
3-chloro-2-fluoro-5-trifluoromethyl pyridine, 2-fluoro-3-chloro-5-trifluoromethyl pyridine, 2-fluoro-3-chloro-5-trifluoromethylpyridine, pyridine, 3-chloro-2-fluoro-5-trifluoromethyl, acmc-20aied, librarion l939, 3-chloro-2-fluoro-5 trifluoromethyl pyridine, 3-chloro-2-fluoro-5-trifluoromethyl-pyridine
1268075
3-chloro-2-fluoro-5-(trifluoromethyl)pyridine
FC1=C(Cl)C=C(C=N1)C(F)(F)F
This item is not returnable. View return policy
72537-17-8
C6H2ClF4N
199.53
MFCD00191918
GDSROTVTTLUHCO-UHFFFAOYSA-N
3-chloro-2-fluoro-5-trifluoromethyl pyridine, 2-fluoro-3-chloro-5-trifluoromethyl pyridine, 2-fluoro-3-chloro-5-trifluoromethylpyridine, pyridine, 3-chloro-2-fluoro-5-trifluoromethyl, acmc-20aied, librarion l939, 3-chloro-2-fluoro-5 trifluoromethyl pyridine, 3-chloro-2-fluoro-5-trifluoromethyl-pyridine
1268075
3-chloro-2-fluoro-5-(trifluoromethyl)pyridine
FC1=C(Cl)C=C(C=N1)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 16°C to 20°C (decomposition)
Color Colorless
Density 1.524 g/mL
Boiling Point 50°C to 55°C (11 mmHg)
Flash Point 52°C (126°F)
Refractive Index 1.433
Quantity 1 g
UN Number UN1993
Formula Weight 199.53
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Chloro-2-fluoro-5-(trifluoromethyl)pyridine
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'