missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloro-2-(chloromethyl)-5-(trifluoromethyl)pyridine, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 95.0 |
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 5 g |
Synonym | 3-chloro-2-chloromethyl-5-trifluoromethyl pyridine, 3-chloranyl-2-chloromethyl-5-trifluoromethyl pyridine, pyridine,3-chloro-2-chloromethyl-5-trifluoromethyl, pyridine, 3-chloro-2-chloromethyl-5-trifluoromethyl |
Formula Weight | 230.02 |
Percent Purity | 95% |
Chemical Name or Material | 3-Chloro-2-(chloromethyl)-5-(trifluoromethyl)pyridine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'