missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2,4-pentanedione, 98%, Thermo Scientific Chemicals

Catalog Number 10636354
Click to view available options
Quantity:
5 mL
100 mL
500 mL
2.5 L
This item is not returnable. View return policy
1694-29-7
C5H7ClO2
134.56
MFCD00009651
VLRGXXKFHVJQOL-UHFFFAOYSA-N
74328
3-chloropentane-2,4-dione
CC(=O)C(C(=O)C)Cl
This item is not returnable. View return policy
1694-29-7
C5H7ClO2
134.56
MFCD00009651
VLRGXXKFHVJQOL-UHFFFAOYSA-N
74328
3-chloropentane-2,4-dione
CC(=O)C(C(=O)C)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point -15°C
Color Brown to Yellow
Density 1.1790g/mL
Boiling Point 49°C to 52°C (18.0 mmHg)
Flash Point 56°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3COCH(Cl)COCH3
Refractive Index 1.4825 to 1.4845
Quantity 500 mL
Beilstein 01, 785
Specific Gravity 1.179
Synonym 3-chloro-2,4-pentanedione, 3-chloroacetylacetone, 2,4-pentanedione, 3-chloro, 1-acetyl-1-chloroacetone, 3-chloro 2,4-pentanedione, 3-chloro.2,4-pentanedione, 3-chloro-pentane-2,4-dione, 2,4-pentanedione,3-chloro, 3-chloro-2,4-pentanedione, produced by wacker chemie ag, burghausen, germany gc
Solubility Information Solubility in water: 7.65g/L (20°C). Other solubilities: soluble in organic solvents
Formula Weight 134.56
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Chloro-2, 4-pentanedione, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Chloro-2,4-pentanedione
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'