missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Chloro-2,4-difluorobenzonitrile, 95%

Brand:  Alfa Aesar™ H63753.03

Code : NEW

Additional Details : CAS Number : 887267-38-1 Weight : 0.00100kg Transport : UN number : 3439 Chem class : 1 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 15410609

  • £68.90 / 1g


Estimated Shipment date
from Supplier 13-02-2018
Add to basket

Specifications

Chemical Identifiers

3-Chloro-2,4-difluorobenzonitrile
173.55
UN3439
887267-38-1
MFCD06658267

Specifications

3-Chloro-2,4-difluorobenzonitrile
1g
Powder
173.55
UN3439
Off-white
887267-38-1
95%
MFCD06658267
Videos
SDS