missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-1-methyl-1H-pyrazole, 97%, Thermo Scientific™

Catalog Number p-3586718
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
63425-54-7
C4H5ClN2
116.55
MFCD09879994
GWZMWAZUCSGVLQ-UHFFFAOYSA-N
3-chloro-1-methyl-1h-pyrazole, 1h-pyrazole, 3-chloro-1-methyl
13630086
3-chloro-1-methylpyrazole
CN1C=CC(Cl)=N1
This item is not returnable. View return policy
63425-54-7
C4H5ClN2
116.55
MFCD09879994
GWZMWAZUCSGVLQ-UHFFFAOYSA-N
3-chloro-1-methyl-1h-pyrazole, 1h-pyrazole, 3-chloro-1-methyl
13630086
3-chloro-1-methylpyrazole
CN1C=CC(Cl)=N1
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 116.55
Percent Purity 97%
Chemical Name or Material 3-Chloro-1-methyl-1H-pyrazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'