missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-3-Chloro-1,2-propanediol, 98%, Thermo Scientific Chemicals

Catalog Number 11309509
Click to view available options
Quantity:
10,000 g
250 g
2500 g
500 g
This item is not returnable. View return policy
96-24-2
C3H7ClO2
110.54
MFCD00004712
SSZWWUDQMAHNAQ-UHFFFAOYNA-N
3-chloro-1,2-propanediol, alpha-chlorohydrin, chlorodeoxyglycerol, glycerol alpha-monochlorohydrin, epibloc, 1,2-propanediol, 3-chloro, a-chlorohydrin, 3-chloropropanediol, 3-chloropropylene glycol, glycerol chlorohydrin
7290
CHEBI:18721
3-chloropropane-1,2-diol
OCC(O)CCl
This item is not returnable. View return policy
96-24-2
C3H7ClO2
110.54
MFCD00004712
SSZWWUDQMAHNAQ-UHFFFAOYNA-N
3-chloro-1,2-propanediol, alpha-chlorohydrin, chlorodeoxyglycerol, glycerol alpha-monochlorohydrin, epibloc, 1,2-propanediol, 3-chloro, a-chlorohydrin, 3-chloropropanediol, 3-chloropropylene glycol, glycerol chlorohydrin
7290
CHEBI:18721
3-chloropropane-1,2-diol
OCC(O)CCl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -40°C
Boiling Point 213°C
Flash Point 137°C (278°F)
Quantity 250 g
UN Number UN2689
Beilstein 635684
Merck Index 14,2145
Formula Weight 110.54
Percent Purity 98%
Chemical Name or Material (+/-)-3-Chloro-1,2-propanediol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'