missing translation for 'onlineSavingsMsg'
Learn More

3-chloro-1,2-benzisoxazole, 97%, Thermo Scientific™

Catalog Number p-2699980
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
16263-52-8
C7H4ClNO
153.565
MFCD03407314
INWUFXPCLZRSBH-UHFFFAOYSA-N
2779780
3-chloro-1,2-benzoxazole
C1=CC=C2C(=C1)C(=NO2)Cl
This item is not returnable. View return policy
16263-52-8
C7H4ClNO
153.565
MFCD03407314
INWUFXPCLZRSBH-UHFFFAOYSA-N
2779780
3-chloro-1,2-benzoxazole
C1=CC=C2C(=C1)C(=NO2)Cl
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 3-chloro-1,2-benzisoxazole, 3-chlorobenzo d isoxazole, 1,2-benzisoxazole,3-chloro, 3-chloro-1,2-benzoisoxazole, 3-chloro-1,2-benzisooxazole, 3-chloranyl-1,2-benzoxazole, 3-chlorobenzisoxazole, pubchem8716, zlchem 1012, 1,2-benzisoxazole, 3-chloro
Formula Weight 153.57
Percent Purity 97%
Chemical Name or Material 3-chloro-1, 2-benzisoxazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'