missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-chloro-1,2-benzisoxazole, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Synonym | 3-chloro-1,2-benzisoxazole, 3-chlorobenzo d isoxazole, 1,2-benzisoxazole,3-chloro, 3-chloro-1,2-benzoisoxazole, 3-chloro-1,2-benzisooxazole, 3-chloranyl-1,2-benzoxazole, 3-chlorobenzisoxazole, pubchem8716, zlchem 1012, 1,2-benzisoxazole, 3-chloro |
Formula Weight | 153.57 |
Percent Purity | 97% |
Chemical Name or Material | 3-chloro-1, 2-benzisoxazole |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'