missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Carboxy-5-fluorobenzeneboronic acid, 98%

Catalog Number p-7035172
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
871329-84-9
C7H6BFO4
183.929
MFCD07363791
APHPXZJIUTVQNC-UHFFFAOYSA-N
44717205
3-borono-5-fluorobenzoic acid
B(C1=CC(=CC(=C1)F)C(=O)O)(O)O
This item is not returnable. View return policy
871329-84-9
C7H6BFO4
183.929
MFCD07363791
APHPXZJIUTVQNC-UHFFFAOYSA-N
44717205
3-borono-5-fluorobenzoic acid
B(C1=CC(=CC(=C1)F)C(=O)O)(O)O
Melting Point 264°C to 266°C
Quantity 250 mg
Synonym 3-carboxy-5-fluorophenylboronic acid, 3-carboxy-5-fluoro benzeneboronic acid, 3-dihydroxyboranyl-5-fluorobenzoic acid, acmc-209qgr, 3-borono-5-fluorobenzoicacid, ksc657s3f, 3-borono-5-fluoro-benzoic acid, benzoic acid,3-borono-5-fluoro, benzoic acid, 3-borono-5-fluoro, 3-carboxy-5-fluorobenzeneboronic acid
Formula Weight 183.93
Percent Purity 98%
Chemical Name or Material 3-Carboxy-5-fluorobenzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'