missing translation for 'onlineSavingsMsg'
Learn More

3-Carbethoxy-4-piperidone hydrochloride, 98%, Thermo Scientific Chemicals

Catalog Number 11976281
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
4644-61-5
C8H13NO3·HCl
207.66
MFCD00043267
RTYXWLMQWSFXNI-UHFFFAOYSA-N
ethyl 4-piperidone-3-carboxylate hydrochloride, ethyl 4-oxopiperidine-3-carboxylate hydrochloride, 3-carbethoxy-4-piperidone hydrochloride, ethyl 4-oxo-3-piperidinecarboxylate hydrochloride, 3-carbethoxy-4-piperidone hcl, 3-ethoxycarbonyl-4-piperidone hydrochloride, ethyl 4-piperidone-3-carboxylate hcl, ethyl 4-piperidone-3-carboxylate, hcl, 3-piperidinecarboxylic acid, 4-oxo-, ethyl ester, hydrochloride, 4-oxo-piperidine-3-carboxylic acid ethyl ester hydrochloride
107315
ethyl 4-oxopiperidine-3-carboxylate;hydrochloride
CCOC(=O)C1CNCCC1=O.Cl
This item is not returnable. View return policy
4644-61-5
C8H13NO3·HCl
207.66
MFCD00043267
RTYXWLMQWSFXNI-UHFFFAOYSA-N
ethyl 4-piperidone-3-carboxylate hydrochloride, ethyl 4-oxopiperidine-3-carboxylate hydrochloride, 3-carbethoxy-4-piperidone hydrochloride, ethyl 4-oxo-3-piperidinecarboxylate hydrochloride, 3-carbethoxy-4-piperidone hcl, 3-ethoxycarbonyl-4-piperidone hydrochloride, ethyl 4-piperidone-3-carboxylate hcl, ethyl 4-piperidone-3-carboxylate, hcl, 3-piperidinecarboxylic acid, 4-oxo-, ethyl ester, hydrochloride, 4-oxo-piperidine-3-carboxylic acid ethyl ester hydrochloride
107315
ethyl 4-oxopiperidine-3-carboxylate;hydrochloride
CCOC(=O)C1CNCCC1=O.Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 169.0°C to 175.0°C
Assay Percent Range 98%
Quantity 100 g
Formula Weight 207.66
Percent Purity 98%
Chemical Name or Material 3-Carbethoxy-4-piperidone hydrochloride
compliance-icons
Product Identifier
  • 3-Carbethoxy-4-piperidone hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'