missing translation for 'onlineSavingsMsg'
Learn More

3-Butyn-2-one, 96%, Thermo Scientific Chemicals

Catalog Number p-4510577
Click to view available options
Quantity:
2.5 g
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1423-60-5
C4H4O
68.08
MFCD00008775
XRGPFNGLRSIPSA-UHFFFAOYSA-N
3-butyn-2-one, 1-butyn-3-one, methyl ethynyl ketone, acetylacetylene, butyn-2-one, acetylethyne, 1-butyne-3-one, 3-butyne-2-one, ethynyl methyl ketone, ch3coc#ch
15018
CHEBI:48060
but-3-yn-2-one
CC(=O)C#C
This item is not returnable. View return policy
1423-60-5
C4H4O
68.08
MFCD00008775
XRGPFNGLRSIPSA-UHFFFAOYSA-N
3-butyn-2-one, 1-butyn-3-one, methyl ethynyl ketone, acetylacetylene, butyn-2-one, acetylethyne, 1-butyne-3-one, 3-butyne-2-one, ethynyl methyl ketone, ch3coc#ch
15018
CHEBI:48060
but-3-yn-2-one
CC(=O)C#C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 0.8700g/mL
Boiling Point 85.0°C
Flash Point −1°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula HC≡CCOCH3
Refractive Index 1.4045 to 1.4095
Quantity 2.5 g
Fieser 03,38
Specific Gravity 0.87
Formula Weight 68.06
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 3-Butyn-2-one
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Butyn-2-one
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H300-Fatal if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'