missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-3-Butyn-2-ol, 98%, Thermo Scientific Chemicals
39.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
250 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Density | 0.894 |
Boiling Point | 108°C to 109°C |
Flash Point | 28°C (82°F) |
Refractive Index | 1.426 |
Quantity | 25 g |
UN Number | UN2929 |
Beilstein | 635722 |
Formula Weight | 70.09 |
Percent Purity | 98% |
Chemical Name or Material | (±)-3-Butyn-2-ol |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Self-reactive substance or mixture Type G
- Skin corrosion/irritation Category 2
- H225-Highly flammable liquid and vapour.
- H300-Fatal if swallowed.
- H310-Fatal in contact with skin.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H330-Fatal if inhaled.
- H412-Harmful to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P362+P364-Take off contaminated clothing and wash it before reuse.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'