missing translation for 'onlineSavingsMsg'
Learn More

3-Butyn-2-ol, 97%, Thermo Scientific Chemicals

Catalog Number p-3211875
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2028-63-9
C4H6O
70.09
MFCD00004541
GKPOMITUDGXOSB-UHFFFAOYNA-N
3-butyn-2-ol, 3-butyne-2-ol, 3-hydroxy-1-butyne, 1-butyn-3-ol, 1-methylpropargyl alcohol, methylethynylcarbinol, 1-ethynylethanol, 1-methyl-2-propynyl alcohol, alpha-methylpropargyl alcohol, dl-3-butyn-2-ol
16239
but-3-yn-2-ol
CC(O)C#C
This item is not returnable. View return policy
2028-63-9
C4H6O
70.09
MFCD00004541
GKPOMITUDGXOSB-UHFFFAOYNA-N
3-butyn-2-ol, 3-butyne-2-ol, 3-hydroxy-1-butyne, 1-butyn-3-ol, 1-methylpropargyl alcohol, methylethynylcarbinol, 1-ethynylethanol, 1-methyl-2-propynyl alcohol, alpha-methylpropargyl alcohol, dl-3-butyn-2-ol
16239
but-3-yn-2-ol
CC(O)C#C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8900g/mL
Boiling Point 108°C to 111°C
Flash Point 26°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4250 to 1.427
Quantity 10 g
Specific Gravity 0.89
Solubility Information Solubility in water: miscible. Other solubilities: miscible with methanol
Formula Weight 70.09
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Butyn-2-ol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 3-Butyn-2-ol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Self-reactive substance or mixture Type G
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P362+P364-Take off contaminated clothing and wash it before reuse.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'