missing translation for 'onlineSavingsMsg'
Learn More

3-Bromopropionic acid, 97%, Thermo Scientific Chemicals

Catalog Number 10657694
Click to view available options
Quantity:
10 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
590-92-1
C3H5BrO2
152.98
MFCD00002763
DHXNZYCXMFBMHE-UHFFFAOYSA-N
3-bromopropionic acid, propanoic acid, 3-bromo, beta-bromopropionic acid, propionic acid, 3-bromo, 2-carboxyethyl bromide, beta-bromopropanoic acid, unii-wfz7csr69r, beta-propionic acid, 3-bromo-propionic acid, wfz7csr69r
11553
3-bromopropanoic acid
OC(=O)CCBr
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
590-92-1
C3H5BrO2
152.98
MFCD00002763
DHXNZYCXMFBMHE-UHFFFAOYSA-N
3-bromopropionic acid, propanoic acid, 3-bromo, beta-bromopropionic acid, propionic acid, 3-bromo, 2-carboxyethyl bromide, beta-bromopropanoic acid, unii-wfz7csr69r, beta-propionic acid, 3-bromo-propionic acid, wfz7csr69r
11553
3-bromopropanoic acid
OC(=O)CCBr

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 57°C to 63°C
Color White to Yellow
Boiling Point 140°C to 142°C (45.0 mmHg)
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Plastic bottle
Linear Formula BrCH2CH2CO2H
Quantity 500 g
Beilstein 02, 256
Merck Index 15, 1444
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol,ether,chcl3 and benzene
Formula Weight 152.98
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 3-Bromopropionic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'