missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(Bromomethyl)thiophene, 97%

Brand:  Alfa Aesar™ H61497.03

Code : NEW

Additional Details : CAS Number : 34846-44-1 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15498048

  • £54.50 / 1g


Estimated Shipment date
from Supplier 24-05-2018
Add to basket

Specifications

Chemical Identifiers

3-(Bromomethyl)thiophene
177.06
106459
34846-44-1
MFCD03840497

Specifications

3-(Bromomethyl)thiophene
1.6030
76°C (1mmHg)
Liquid
97%
MFCD03840497
34846-44-1
1.616g/mL
Pale brown
1g
177.06
106459
Videos
SDS