missing translation for 'onlineSavingsMsg'
Learn More

3-(Bromomethyl)thiophene, 97%, Thermo Scientific Chemicals

Catalog Number p-7053809
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
34846-44-1
C5H5BrS
177.06
MFCD03840497
KBWHYRUAHXHHFO-UHFFFAOYSA-N
3-bromomethyl thiophene, 3-bromomethyl-thiophene, 3-thenyl bromide, thiophene, 3-bromomethyl, 3-thienylmethyl bromide, thien-3-ylmethyl bromide, zlchem 651, 3-brommethyl-thiophen, pubchem5295, 3 bromomethyl thiophene
520703
3-(bromomethyl)thiophene
BrCC1=CSC=C1
This item is not returnable. View return policy
34846-44-1
C5H5BrS
177.06
MFCD03840497
KBWHYRUAHXHHFO-UHFFFAOYSA-N
3-bromomethyl thiophene, 3-bromomethyl-thiophene, 3-thenyl bromide, thiophene, 3-bromomethyl, 3-thienylmethyl bromide, thien-3-ylmethyl bromide, zlchem 651, 3-brommethyl-thiophen, pubchem5295, 3 bromomethyl thiophene
520703
3-(bromomethyl)thiophene
BrCC1=CSC=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Brown
Density 1.616 g/mL
Boiling Point 76°C (1 mmHg)
Refractive Index 1.603
Quantity 1 g
Beilstein 106459
Sensitivity Moisture sensitive
Formula Weight 177.06
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-(Bromomethyl)thiophene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'