missing translation for 'onlineSavingsMsg'
Learn More

3-(Bromomethyl)benzaldehyde, 97%, Maybridge

Brand:  Maybridge MO07388DE

Code : 50

Additional Details : CAS Number : 82072-23-9 Weight : 0.05500kg Transport : UN number : 3261 Chem class : 8 Pack group : III

Quantity 5g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10664205

  • £301.00 / 5g


Estimated Shipment date
from Supplier 23-05-2018
Add to basket

Specifications

Chemical Identifiers

3-(Bromomethyl)benzaldehyde
97%
199.05
97%
C8H7BrO

Specifications

3-(Bromomethyl)benzaldehyde
Amber Glass Bottle
97%
C8H7BrO
82072-23-9
5g
97%
199.05
Videos
SDS