missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromofuran, 97%, stab. with 0.5% calcium carbonate, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
3-Bromofuran was a synthesis reagent used in the preparation of 2-substituted 3-furfurals. It was also used in the synthesis of 5,6-dehydronorcantharidins. It is one of the volatile metabolites discussed in studies for detecting and discriminating diseases of onion.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Bromofuran was a synthesis reagent used in the preparation of 2-substituted 3-furfurals. It was also used in the synthesis of 5,6-dehydronorcantharidins. It is one of the volatile metabolites discussed in studies for detecting and discriminating diseases of onion.
Solubility
Soluble in chloroform, ethyl acetate. Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from heat, oxidizing agents.
3-Bromofuran was a synthesis reagent used in the preparation of 2-substituted 3-furfurals. It was also used in the synthesis of 5,6-dehydronorcantharidins. It is one of the volatile metabolites discussed in studies for detecting and discriminating diseases of onion.
Solubility
Soluble in chloroform, ethyl acetate. Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from heat, oxidizing agents.
Specifications
Density | 1.635 |
Boiling Point | 102°C to 104°C |
Flash Point | 3°C (37°F) |
Refractive Index | 1.496 |
Quantity | 1 g |
UN Number | UN1993 |
Beilstein | 105337 |
Solubility Information | Soluble in chloroform,ethyl acetate. Insoluble in water. |
Formula Weight | 146.98 |
Percent Purity | 97% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'