missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromofuran, 97%, Thermo Scientific™
Click to view available options
Quantity:
5 g
10 g
25 g
Specifications
CAS Min % | 94.5, 0.05, 0.45, 0.35 |
CAS Max % | 98.5, 0.15, 0.55, 0.45 |
Density | 1.635g/mL |
Assay Percent Range | 95% |
Quantity | 5 g |
Formula Weight | 146.97 |
Percent Purity | 95% |
Chemical Name or Material | 3-Bromofuran, stabilized with Cu(0.1%), Furan (0.5%) & NaHCO3 (0.4%) |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'