missing translation for 'onlineSavingsMsg'
Learn More

3-Bromofuran, 97%, Thermo Scientific™

Catalog Number 10501773
Click to view available options
Quantity:
5 g
10 g
25 g
This item is not returnable. View return policy
22037-28-1, 7440-50-8, 110-00-9, 144-55-8
C4H3BrO
146.97
MFCD00005347
LXWLEQZDXOQZGW-UHFFFAOYSA-N
furan, 3-bromo, .beta.-bromofuran, beta-bromofuran, 3-furyl bromide, 3-bromo furan, 3-bromo-furan, zlchem 308, pubchem8830, 3-bromofuran, acmc-209fqz
89164
3-bromofuran
BrC1=COC=C1
This item is not returnable. View return policy
22037-28-1, 7440-50-8, 110-00-9, 144-55-8
C4H3BrO
146.97
MFCD00005347
LXWLEQZDXOQZGW-UHFFFAOYSA-N
furan, 3-bromo, .beta.-bromofuran, beta-bromofuran, 3-furyl bromide, 3-bromo furan, 3-bromo-furan, zlchem 308, pubchem8830, 3-bromofuran, acmc-209fqz
89164
3-bromofuran
BrC1=COC=C1
CAS Min % 94.5, 0.05, 0.45, 0.35
CAS Max % 98.5, 0.15, 0.55, 0.45
Density 1.635g/mL
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 10 g
Formula Weight 146.97
Percent Purity 95%
Chemical Name or Material 3-Bromofuran, stabilized with Cu(0.1%), Furan (0.5%) & NaHCO3 (0.4%)
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'