missing translation for 'onlineSavingsMsg'
Learn More

3-Bromofluorobenzene, 99%, Thermo Scientific Chemicals

Catalog Number p-74613
Click to view available options
Quantity:
25 mL
100 mL
500 mL
This item is not returnable. View return policy
1073-06-9
C6H4BrF
175
MFCD00000326
QDFKKJYEIFBEFC-UHFFFAOYSA-N
3-bromofluorobenzene, m-bromofluorobenzene, m-fluorobromobenzene, benzene, 1-bromo-3-fluoro, 3-bromfluorbenzen, 3-fluorobromobenzene, m-fluorophenyl bromide, 1-fluoro-3-bromobenzene, 3-fluoro bromobenzene, benzene, 1-fluoro-3-bromo
14082
1-bromo-3-fluorobenzene
C1=CC(=CC(=C1)Br)F
This item is not returnable. View return policy
1073-06-9
C6H4BrF
175
MFCD00000326
QDFKKJYEIFBEFC-UHFFFAOYSA-N
3-bromofluorobenzene, m-bromofluorobenzene, m-fluorobromobenzene, benzene, 1-bromo-3-fluoro, 3-bromfluorbenzen, 3-fluorobromobenzene, m-fluorophenyl bromide, 1-fluoro-3-bromobenzene, 3-fluoro bromobenzene, benzene, 1-fluoro-3-bromo
14082
1-bromo-3-fluorobenzene
C1=CC(=CC(=C1)Br)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.5670g/mL
Boiling Point 150°C
Flash Point 38°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Linear Formula BrC6H4F
Refractive Index 1.5260 to 1.528
Quantity 25 mL
Specific Gravity 1.567
Solubility Information Solubility in water: insoluble in water. Other solubilities: organic soluble/miscible with most solvents
Formula Weight 175
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3-Bromofluorobenzene, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Bromofluorobenzene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'